Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Il3 (P01586, 33 a.a. - 166 a.a.) Partial Recombinant Protein

Catalog No. 89948877
Click to view available options
Quantity:
10 μg

Used for Func, SDS-PAGE

Sequence: DTHRLTRTLNCSSIVKEIIGKLPEPELKTDDEGPSLRNKSFRRVNLSKFVESQGEVDPEDRYVIKSNLQKLNCCLPTSANDSALPGVFIRDLDDFRKKLRFYMVHLNDLETVLTSRPPQPASGSVSPNRGTVEC

Specifications

Accession Number P01586
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 16187
Molecular Weight (g/mol) 15kDa
Name Il3 (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 10 μg
Immunogen DTHRLTRTLNCSSIVKEIIGKLPEPELKTDDEGPSLRNKSFRRVNLSKFVESQGEVDPEDRYVIKSNLQKLNCCLPTSANDSALPGVFIRDLDDFRKKLRFYMVHLNDLETVLTSRPPQPASGSVSPNRGTVEC
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1EU/μg) (determined by LAL gel clot method)
Gene Alias BPA/Csfmu/HCGF/Il-3/MCGF/PSF
Common Name Il3
Gene Symbol Il3
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of mouse M-NFS-60 cells was found to be in the < 0.05ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.