Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Indian Hedgehog/Ihh Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB8048
This item is not returnable.
View return policy
Description
Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Mouse samples. It is validated for Neutralization.Specifications
| Indian Hedgehog/Ihh | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDA1, BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Ihh, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3549 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization | |
| 852526 | |
| Neutralization 10-45 ug/mL | |
| P97812 | |
| IHH | |
| E. coli-derived recombinant mouse Indian Hedgehog/Ihh Tyr67-Phe240 Accession # P97812 | |
| 500 μg | |
| Primary | |
| Detects mouse Indian Hedgehog/Ihh in ELISAs. In direct ELISAs, no cross-reactivity with full length recombinant mouse (rm) Dhh or rmShh is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction