Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Indian Hedgehog/Ihh N-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1705
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1705 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1705 Supplier R&D Systems Supplier No. AF1705
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

Indian Hedgehog/Ihh Polyclonal specifically detects Indian Hedgehog/Ihh in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Indian Hedgehog/Ihh
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P97812
Gene Alias BDA1, BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Ihh, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein
Gene Symbols IHH
Host Species Goat
Immunogen E. coli-derived recombinant mouse Indian Hedgehog Cys28-Gly202 Accession # P97812
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3549
Test Specificity Detects mouse Indian Hedgehog (Ihh) N-terminal peptide in Western blots. In Western blots, approximately 80% cross-reactivity with recombinant human Shh N-terminal peptide and recombinant mouse (rm) Shh N-terminal peptide is observed, and less than 1% cross-reactivity with rmIhh C-terminal peptide (amino acids 241-449) is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.