Learn More
Mouse Integrin alpha 10 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Integrin alpha 10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 885501 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1) |
| Gene Alias | integrin alpha-10, integrin, alpha 10, ITGA10, PRO827 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.