Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Integrin alpha 10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9757SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9757SP 25 μg
MAB9757100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9757SP Supplier R&D Systems Supplier No. MAB9757SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin alpha 10 Monoclonal specifically detects Integrin alpha 10 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Integrin alpha 10
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 885501
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
Gene Alias integrin alpha-10, integrin, alpha 10, ITGA10, PRO827
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1 Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1) Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 8515
Test Specificity Detects mouse Integrin alpha 10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Intagrin alpha 2 beta 1 was observered.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.