Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Integrin alpha 3/CD49c PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems FAB2787P
This item is not returnable.
View return policy
Description
Integrin alpha 3/CD49c Polyclonal specifically detects Integrin alpha 3/CD49c in Mouse samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 3/CD49c | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
antigen identified by monoclonal J143, CD49 antigen-like family member C, CD49c, CD49c antigen, FLJ34631, FLJ34704, FRP-2, Galactoprotein B3, GAP-B3, GAPB3CD49C, integrin alpha-3, integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor), ITGA3, MSK18, VCA-2, very late activation protein 3 receptor, alpha-3 subunit, VL3A, VLA-3 subunit alpha, VLA3a | |
Goat | |
Affinity Purified | |
RUO | |
3675 | |
Mouse | |
IgG |
Flow Cytometry | |
R-PE | |
Q62470 | |
ITGA3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Integrin alpha 3/CD49c Phe33-Glu993 Accession # Q62470 | |
100 Tests | |
Primary | |
Detects mouse Integrin alpha 3/CD49c in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Integrin alpha 2, rmIntegrin alpha 4, rmIntegrin alpha 5, rmIntegrin alpha E, rmIntegrin alpha M, rmIntegrin alpha V and rmIntegrin alpha X is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction