Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Integrin alpha 7 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3518T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3518T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3518T100 Supplier R&D Systems Supplier No. FAB3518T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin alpha 7 Monoclonal specifically detects Integrin alpha 7 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 7
Applications Flow Cytometry
Classification Monoclonal
Clone 334908
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_032424
Gene Alias FLJ25220, integrin alpha 7 chain, integrin alpha-7, integrin, alpha 7, ITGA7
Gene Symbols ITGA7
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 7 Phe34-Pro1036 Accession # NP_032424
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3679
Test Specificity Detects mouse Integrin alpha 7 in direct ELISAs. Reactivity with other Integrin alpha 7 isoforms was not tested. In direct ELISAs, no cross-reactivity with recombinant mouse Integrin alpha 2, alpha 3, alpha 4, alpha 5, alpha 9, alpha E, alpha X, recombinant human Integrin alpha L, or alpha 11 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.