Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Integrin alpha 7 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB3518SP
This item is not returnable.
View return policy
Description
Integrin alpha 7 Monoclonal specifically detects Integrin alpha 7 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Integrin alpha 7 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
NP_032424 | |
ITGA7 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 7 Phe34-Pro1036 Accession # NP_032424 | |
25 μg | |
Primary | |
Detects mouse Integrin alpha 7 in direct ELISAs. Reactivity with other Integrin alpha 7 isoforms was not tested. In direct ELISAs, no cross-reactivity with recombinant mouse Integrin alpha 2, alpha 3, alpha 4, alpha 5, alpha 9, alpha E, alpha X, recombinant human Integrin alpha L, or alpha 11 is observed. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
334908 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ25220, integrin alpha 7 chain, integrin alpha-7, integrin, alpha 7, ITGA7 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3679 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction