Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Integrin alpha 9 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3827P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3827P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3827P Supplier R&D Systems Supplier No. FAB3827P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Integrin alpha 9 Polyclonal specifically detects Integrin alpha 9 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 9
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. NP_598482
Gene Alias ALPHA-RLC, integrin alpha-9, Integrin alpha-RLC, integrin, alpha 9, ITGA4L, ITGA9, RLC, RLC-alpha
Gene Symbols ITGA9
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Integin alpha 9 Try31-Val979 Accession # NP_598482
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3680
Test Specificity Detects mouse Integrin alpha 9 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human Integrin alpha 1, alpha 8, alpha 9, alpha L, recombinant mouse Integrin 3, 4, 5, E, and M is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.