Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Integrin alpha E/CD103 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1990
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1990 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1990 Supplier R&D Systems Supplier No. BAF1990
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Integrin alpha E/CD103 Polyclonal specifically detects Integrin alpha E/CD103 in Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Integrin alpha E/CD103
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells
Gene Accession No. ABD49099
Gene Alias antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), ITGAE, MGC141996, Mucosal lymphocyte 1 antigen
Gene Symbols ITGAE
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha E Phe20-Arg1113 (Ser453Gly) Accession # ABD49099
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3682
Test Specificity Detects mouse Integrin alpha E/CD103 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Integrin alpha 2, rmIntegrin alpha 3, rmIntegrin alpha 4, rmIntegrin alpha M, and recombinant human Integrin alpha V is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.