Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Integrin beta 1/CD29 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2405N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2405N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2405N100 Supplier R&D Systems Supplier No. FAB2405N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin beta 1/CD29
Applications Flow Cytometry
Classification Monoclonal
Clone 265917
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P09055
Gene Alias CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), ITGB1, MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Gene Symbols ITGB1
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 1/CD29 Gln21-Asp728 Accession # P09055
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3688
Test Specificity Detects mouse Integrin beta 1/CD29 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Integrin beta 1, recombinant mouse (rm) Integrin beta 2, rmIntegrin beta 3, rmIntegrin beta 4, or rmIntegrin beta 6 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.