Learn More
Mouse Integrin beta 1/CD29 PE-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Integrin beta 1/CD29 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 265917 |
| Conjugate | R-PE |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | P09055 |
| Gene Alias | CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), ITGB1, MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA |
| Gene Symbols | ITGB1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.