Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Integrin beta 3/CD61 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA41182R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA41182R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA41182R100 Supplier R&D Systems Supplier No. FAB41182R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin beta 3/CD61
Applications Flow Cytometry
Classification Monoclonal
Clone 909114
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_034705 (Integrin alpha 2b) & O54890 (Integrin beta 3)
Gene Alias CD61, CD61 antigen, GP3A, GP3Aplatelet glycoprotein IIIa, GPIIIA, GPIIIaintegrin beta-3, INGRB3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, ITGB3, Platelet membrane glycoprotein IIIa
Gene Symbols ITGB3
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 2b and Integrin beta 3 linked heterodimer Leu32-Arg988 (Integrin alpha 2b) & Glu26-Asp717 (Integrin beta 3) Accession # NP_034705 (Integrin alpha 2b) & O54890 (Integrin beta 3)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3690
Test Specificity Detects mouse Integrin beta 3/CD61 in direct ELISA.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.