Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Integrin beta 6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2389
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2389 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2389 Supplier R&D Systems Supplier No. BAF2389
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Integrin beta 6 Polyclonal specifically detects Integrin beta 6 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Integrin beta 6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q9Z0T9
Gene Alias integrin beta-6, integrin, beta 6, ITGB6
Gene Symbols ITGB6
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 6 Gly22-Asn706 Accession # Q9Z0T9
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3694
Test Specificity Detects mouse Integrin beta 6 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse (rm) Integrin beta 1, rmIntegrin beta 2, and rmIntegrin beta 3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.