Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Intelectin-1/Omentin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8074
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8074 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8074 Supplier R&D Systems Supplier No. MAB8074
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Intelectin-1/Omentin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 746420
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O88310
Gene Alias Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, Intelectin1, INTLHL1, ITLN1, Itlna, ITLNITLN-1, LFR, LFRIntestinal lactoferrin receptor, omentin, Omentin-1
Gene Symbols ITLN1
Host Species Rat
Immunogen E. coli-derived recombinant mouse Intelectin-1/Omentin Ala20-Ser298 Accession # O88310
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55600
Test Specificity Detects mouse Intelectin-1/Omentin in ELISA. In direct ELISAs, no significant cross-reactivity with recombinant mouse Intelectin-2 and recombinant human Intelectin-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.