Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Jagged 2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB4748R100UG
This item is not returnable.
View return policy
Description
Jagged 2 Monoclonal specifically detects Jagged 2 in Mouse samples. It is validated for Flow Cytometry.Specifications
Jagged 2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9QYE5 | |
JAG2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Jagged 2 Met27-Leu1084 Accession # Q9QYE5 | |
RUO | |
3714 | |
Mouse | |
IgG2a |
Flow Cytometry | |
746513 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
HJ2, JAG2, jagged 2, Jagged2, protein jagged-2, SER2 | |
Rat | |
100 μg | |
Primary | |
Detects mouse Jagged 2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Jagged 1, rhJagged 2, recombinant rat Jagged 1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction