Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse JAM-A Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1077
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1077 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1077 Supplier R&D Systems Supplier No. BAF1077
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

JAM-A Polyclonal specifically detects JAM-A in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen JAM-A
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. O88792
Gene Alias CD321, CD321 antigen, F11 receptor, F11R, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1, PAM-1KAT, Platelet adhesion molecule 1
Gene Symbols F11R
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse JAM-A Lys27-Ala242 Accession # O88792
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50848
Test Specificity Detects mouse JAM-A in ELISAs in Western blots. In sandwich immunoassays, less than 1% cross-reactivity is observed with recombinant human JAM-A, recombinant mouse (rm) JAM-B, and rmJAM-C.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.