Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse JAM-B/VE-JAM Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB9881100
This item is not returnable.
View return policy
Description
JAM-B/VE-JAM Monoclonal specifically detects JAM-B/VE-JAM in Mouse samples. It is validated for Neutralization.Specifications
JAM-B/VE-JAM | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C21orf43, CD322, CD322 antigen, JAM2, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAM, VE-JAMVEJAMCD322 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
58494 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Neutralization | |
150005 | |
Neutralization 0.1-0.5 ug/mL | |
Q9JI59 | |
JAM2 | |
Mouse myeloma cell line NS0-derived recombinant mouse JAM-B/VE-JAM Phe29-Asn236 (predicted) Accession # Q9JI59 | |
100 μg | |
Primary | |
Detects mouse JAM-B/VE-JAM in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human JAM-1, -2, or -3 is observed. | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction