Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Kallikrein 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7928
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7928 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7928 Supplier R&D Systems Supplier No. AF7928
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Kallikrein 1 Polyclonal specifically detects Kallikrein 1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Kallikrein 1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_081082, P15947
Gene Alias EC 3.4.21, EC 3.4.21.35, glandular kallikrein 1, hK1, kallikrein 1, kallikrein 1, renal/pancreas/salivary, kallikrein serine protease 1, kallikrein-1, Kidney/pancreas/salivary gland kallikrein, KLK1, Klk6, KLKR, Tissue kallikrein
Gene Symbols KLK1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Kallikrein 5 precursor/Kallikrein 1 Val23-Arg67(Kallikrein 5 precursor), Ile25-Asp261(Kallikrein 1) Accession # NP_081082, P15947
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3816
Test Specificity Detects mouse Kallikrein 1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Kallikrein 1, recombinant mouse (rm) Kallikrein 5 and rmKallikrein 7 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.