Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Kininogen Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2206SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2206SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2206SP Supplier R&D Systems Supplier No. MAB2206SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Kininogen Monoclonal specifically detects Kininogen in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Kininogen
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 288422
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O08677
Gene Symbols Kng1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Kininogen Glu21-Ser661 Accession # O08677
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3827
Test Specificity Detects mouse Kininogen in direct ELISAs and Western blots. In Western blots, reacts with the full length mouse Kininogen (aa 21-661), but not with the light chain alone. In Western blots, no cross-reactivity with recombinant human (rh) Cystatin D, F, SA, SN, recombinant mouse (rm) Cystatin A, B, C, E/M, rmCystatin/Stefin Homolog, rmFetuin A, rhFetuin B, rmHPRG, rhKininogen, or rhKininostatin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.