Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Kitl (P20826, 36 a.a. - 189 a.a.) Partial Recombinant Protein expressed in Insect cells

Catalog No. 89949954 Shop All Abnova Corporation Products
Click to view available options
Quantity:
10 μg

Used for Func, SDS-PAGE

Sequence: KEICGNPVTDNVKDITKLVANLPNDYMITLNYVAGMDVLPSHCWLRDMVIQLSLSLTTLLDKFSNISEGLSNYSIIDKLGKIVDDLVLCMEENAPKNIKESPKRPETRSFTPEEFFSIFNRSIDAFKDFMVASDTSDCVLSSTLGPEKDSRVSVTKPFMLPPVA

Specifications

Accession Number P20826
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 17311
Molecular Weight (g/mol) 18kDa
Name Kitl (Mouse) Recombinant Protein
Preparation Method Insect cell expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 10 μg
Immunogen KEICGNPVTDNVKDITKLVANLPNDYMITLNYVAGMDVLPSHCWLRDMVIQLSLSLTTLLDKFSNISEGLSNYSIIDKLGKIVDDLVLCMEENAPKNIKESPKRPETRSFTPEEFFSIFNRSIDAFKDFMVASDTSDCVLSSTLGPEKDSRVSVTKPFMLPPVA
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1EU/μg) (determined by LAL gel clot method)
Gene Alias Clo/Con/Gb/Kitlg/Mgf/SCF/SF/SLF/Sl/Steel/contrasted
Common Name Kitl
Gene Symbol Kitl
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is < 2.0ng/mL, corresponding to a specific activity of ≥ 5 x 105 units/mg.
Species Baculovirus
Recombinant Recombinant
Protein Tag None
Expression System Insect cell expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.