Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse KLF4 Antibody, R&D Systems™



Goat Polyclonal Antibody has been used in 38 publications
Supplier: R&D Systems AF3158
This item is not returnable.
View return policy
Description
KLF4 Polyclonal specifically detects KLF4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.Specifications
KLF4 | |
Polyclonal | |
Unconjugated | |
Q60793 | |
KLF4 | |
E. coli-derived recombinant mouse KLF4 Ala11-Phe483 Accession # Q60793 | |
100 μg | |
Primary | |
Detects mouse KLF4 in direct ELISAs and Western blots. | |
Mouse | |
IgG |
ChIP Assay, Immunocytochemistry, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 μg/mL, Immunohistochemistry 1-25 μg/mL, Chromatin Immunoprecipitation (ChIP) 5 μg/5 x 10^6 cells, Immunocytochemistry 5-15 μg/mL | |
Epithelial zinc finger protein EZF, EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut) | |
Goat | |
Affinity Purified | |
RUO | |
9314 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction