Learn More
Specifications
Specifications
| Assay Range | 6.25 - 100 u/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum |
| Specificity | KLH IgM (Mouse) ELISA Kit is for the quantitative measurement of KLH IgM in plasma and serum. |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Mouse |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
