Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Klotho Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $690.00
Specifications
Antigen | Klotho |
---|---|
Clone | 236214 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Description
Klotho Monoclonal specifically detects Klotho in Mouse samples. It is validated for ELISA, Immunohistochemistry.Specifications
Klotho | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
EC 3.2.1, EC 3.2.1.31, KL, klotho | |
KL | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
236214 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
9365 | |
Mouse myeloma cell line NS0-derived recombinant mouse Klotho | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Klotho (aa 23-550 and aa 35-982) in direct ELISAs and Western blots. Does not cross-react with recombinant human Klotho or recombinant mouse Klotho B. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title