Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Klotho beta Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
$227.50 - $596.50
Specifications
Antigen | Klotho beta |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Klotho beta Polyclonal specifically detects Klotho beta in Mouse samples. It is validated for Western Blot.Specifications
Klotho beta | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
KLB | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_112457 | |
152831 | |
Mouse myeloma cell line NS0-derived recombinant mouse Klotho beta (R&D Systems, Catalog # 2619-KB) Phe53-Leu995 Accession # NP_112457 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Klotho beta in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant mouse Klotho is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title