Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Klotho beta Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2619
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2619 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2619 Supplier R&D Systems Supplier No. BAF2619
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Klotho beta Polyclonal specifically detects Klotho beta in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Klotho beta
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_112457
Gene Alias beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213
Gene Symbols KLB
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Klotho beta Phe53-Leu995 Accession # NP_112457
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152831
Test Specificity Detects mouse Klotho beta in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse Klotho is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.