Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Klre-1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB7099T100UG
This item is not returnable.
View return policy
Description
Klre-1 Monoclonal specifically detects Klre-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
Klre-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_705818 | |
Klre1 | |
Chinese hamster ovary cell line, CHO derived recombinant mouse Klre-1 Lys94-Lys226 Accession # NP_705818 | |
RUO | |
3824 | |
Mouse | |
IgG2b |
Flow Cytometry | |
854929 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD94, Klre1, NKG2I | |
Rat | |
100 μg | |
Primary | |
Detects mouse Klre-1 in ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction