Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Klre-1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7099P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7099P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7099P Supplier R&D Systems Supplier No. FAB7099P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Klre-1 Monoclonal specifically detects Klre-1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Klre-1
Applications Flow Cytometry
Classification Monoclonal
Clone 854929
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_705818
Gene Alias CD94, Klre1, NKG2I
Gene Symbols Klre1
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO derived recombinant mouse Klre-1 Lys94-Lys226 Accession # NP_705818
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3824
Test Specificity Detects mouse Klre-1 in ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.