Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse KLRG1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Hamster Monoclonal Antibody
Supplier: R&D Systems FAB69441N100UG
This item is not returnable.
View return policy
Description
KLRG1 Monoclonal specifically detects KLRG1 in Mouse samples. It is validated for Flow Cytometry.Specifications
KLRG1 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
2F1, CLEC15A, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFA, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
IL-2 activated NK cells from C57/BL6 mice Accession # O88713 | |
100 μg | |
10219 | |
Mouse | |
Supernatant |
Flow Cytometry | |
2F1 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O88713 | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse KLRG1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction