Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse KLRG1 Antibody, R&D Systems™

Hamster Monoclonal Antibody
Supplier: R&D Systems MAB69441SP
This item is not returnable.
View return policy
Description
KLRG1 Monoclonal specifically detects KLRG1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
KLRG1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
2F1, CLEC15A, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFA, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
IL-2 activated NK cells from C57/BL6 mice Accession # O88713 | |
25 μg | |
10219 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Flow Cytometry, CyTOF | |
2F1 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
O88713 | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse KLRG1 in direct ELISAs. | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction