Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Kremen-1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1647
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1647 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1647 Supplier R&D Systems Supplier No. BAF1647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Kremen-1 Polyclonal specifically detects Kremen-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen Kremen-1
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. NP_115772
Gene Alias EC 3.4.21, FLJ31863, KREMEM1, KREMEN, kremen protein 1, Kremen1, kringle containing transmembrane protein, kringle containing transmembrane protein 1, Kringle domain-containing transmembrane protein 1, kringle-coding gene marking the eye and the nose, Kringle-containing protein marking the eye and the nose, KRM1, KRM1Dickkopf receptor
Gene Symbols KREMEN1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Kremen-1 Ala20-Gly395 Accession # NP_115772
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83999
Test Specificity Detects mouse Kremen-1 in ELISAs and Western blots. In sandwich immunoassays, approximately 25% cross-reactivity with recombinant human (rh) Kremen-1 is observed and less than 1% cross-reactivity with rhKremen-2 and recombinant mouse Kremen-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.