Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Kynureninase Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7389SP
This item is not returnable.
View return policy
Description
Kynureninase Monoclonal specifically detects Kynureninase in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Kynureninase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q9CXF0 | |
| KYNU | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Kynureninase Met1-Ser464 Accession # Q9CXF0 | |
| 25 μg | |
| Primary | |
| Detects mouse Kynureninase in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Kynureninase is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 771312 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.7.1.3, KYNU, kynureninase, kynureninase (L-kynurenine hydrolase), L-kynurenine hydrolase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8942 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction