Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse L-Selectin/CD62L Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB576SP
This item is not returnable.
View return policy
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
L-Selectin/CD62L | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6402 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Flow Cytometry | |
95205 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
P18337 | |
SELL | |
Mouse myeloma cell line NS0-derived recombinant mouse L-Selectin/CD62L Met1-Asn332 Accession # P18337 | |
25 μg | |
Primary | |
Detects mouse L-Selectin/CD62L in direct ELISAs and Western blots. In Western blots and direct ELISAs, no cross-reactivity with recombinant human L-Selectin, recombinant mouse (rm) P-Selectin, or rmE-Selectin is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction