Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse LAG-3 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA33281T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA33281T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA33281T100 Supplier R&D Systems Supplier No. FAB33281T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

LAG-3 Monoclonal specifically detects LAG-3 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen LAG-3
Applications Flow Cytometry
Classification Monoclonal
Clone 631501
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q61790
Gene Alias CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC
Gene Symbols LAG3
Host Species Rat
Immunogen NS0 mouse myeloma cell line transfected with mouse LAG-3 Ser23-Leu442 Accession # Q61790
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3902
Test Specificity Detects mouse LAG-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LAG-3 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.