Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LAG-3 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB33281N
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal specifically detects LAG-3 in Mouse samples. It is validated for Flow Cytometry.Specifications
LAG-3 | |
Monoclonal | |
Alexa Fluor 700 | |
Q61790 | |
LAG3 | |
NS0 mouse myeloma cell line transfected with mouse LAG-3 Ser23-Leu442 Accession # Q61790 | |
100 Tests | |
3902 | |
Mouse | |
Purified |
Flow Cytometry | |
631501 | |
Flow Cytometry 5 uL/10^6 cells | |
CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse LAG-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LAG-3 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction