Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LAIR1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB100921100
This item is not returnable.
View return policy
Description
LAIR1 Monoclonal specifically detects LAIR1 in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
LAIR1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD305, CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
Mouse myeloma cell line NS0-derived mouse LAIR1, Gln22-Tyr141 & Ser25-Tyr141, Accession # Q8BG84 | |
100 μg | |
3903 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG |
Blocking Assay | |
2459B | |
Blockade of Receptor-ligand Interaction | |
Q8BG84 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects mouse LAIR1 in direct ELISAs. | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction