Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Laminin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB25491SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB25491SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB25491SP Supplier R&D Systems Supplier No. MAB25491SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Laminin-1 Monoclonal specifically detects Laminin-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Laminin-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone AL-3
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias LAMA, LAMA1, laminin subunit alpha 1, Laminin1, Laminin-111, PTBHS, S-LAM-alpha
Host Species Rat
Immunogen Purified fragment of chymotrypsin-digested mouse EHS tumor-derived Laminin-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284217
Test Specificity Detects mouse Laminin-1 in Western blots. This antibody reacts with mouse Laminin-1 beta chain (1).
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.