Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LAP (TGF-beta 1) Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
Supplier: R&D Systems MAB7666
This item is not returnable.
View return policy
Description
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
LAP (TGF-beta 1) | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CED, DPD1, LAP, LAP (TGFbeta 1), TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, transforming growth factor beta 1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7040 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, CyTOF | |
860206 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P04202 | |
TGFB1 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse LAP (TGF-beta 1) Met1-Ser390 Accession # P04202 | |
100 μg | |
Primary | |
Detects mouse LAP (TGF-beta 1) in flow cytometry. | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction