Learn More
Invitrogen™ Mouse TGF beta-1/LAP Uncoated ELISA Kit with Plates

Description
The Mouse Transforming Growth Factor beta 1 (TGFβ1) Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse TGFβ1. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Specifications
Specifications
Accession Number | P04202 |
Assay Range | 0.2 to 12.5 ng/mL |
Assay Sensitivity | 0.2 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Sample Type | Plasma, Serum, Supernatant |
Target Species | Mouse |
For Use With (Application) | ELISA |
For Use With (Equipment) | Colorimetric Microplate Reader |
Gene ID (Entrez) | 21803 |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.