Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Latent TGF-beta bp4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2885
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2885 100 μg
AF2885SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2885 Supplier R&D Systems Supplier No. AF2885
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

Latent TGF-beta bp4 Polyclonal specifically detects Latent TGF-beta bp4 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Latent TGF-beta bp4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAI41006
Gene Alias Latent TGFbeta bp4, latent transforming growth factor beta binding protein 4, LTBP4, LTBP-4, LTBP4L, LTBP4S
Gene Symbols LTBP4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Latent TGF-beta bp4 isoform 1 Ser25-Ala1666 (Arg542-Pro547) Accession # AAI41006
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8425
Test Specificity Detects mouse Latent TGF-beta bp4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) LTBP-1 and rhLTBP-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.