Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LDLR Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2255
This item is not returnable.
View return policy
Description
LDLR Polyclonal specifically detects LDLR in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry.Specifications
| LDLR | |
| Polyclonal | |
| Biotin | |
| Q6GTJ9 | |
| LDLR | |
| Mouse myeloma cell line NS0-derived recombinant mouse LDL R Ala22-Arg790 (Ala23Val, Cys27Gly) Accession # Q6GTJ9 | |
| 50 μg | |
| Primary | |
| Detects mouse LDL R in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human LDL R is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3949 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction