Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Lefty-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$164.60 - $568.49
Specifications
| Antigen | Lefty-1 |
|---|---|
| Clone | 146903 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Lefty-1 Monoclonal specifically detects Lefty-1 in Mouse samples. It is validated for Western Blot.Specifications
| Lefty-1 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AI450052, EBAF, Leftb, lefty, lefty-1, Lefty1, Lefty-B, Stra3, TGFB4 | |
| LEFTY1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 146903 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q64280.1 | |
| 10637 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Lefty-1 Leu136-Pro368 Accession # Q64280.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Lefty-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 10-25% cross-reactivity with recombinant human (rh) Lefty-A and no cross-reactivity with rmArtemin, rhCripto, rdDpp, rhGDNF, rrGDNF, rhLAP, rrMIS, rhNeurturin, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, or rhTGF-beta 3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title