Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Lefty-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7648SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7648SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7648SP Supplier R&D Systems Supplier No. MAB7648SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Lefty-2 Monoclonal specifically detects Lefty-2 in Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen Lefty-2
Applications Immunocytochemistry
Classification Monoclonal
Clone 836101
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P57785
Gene Alias EBAF, Endometrial Bleeding-Associated Factor, LEFTA, Left-Right Determination Factor 2, Left-right determination factor A, LEFTYA, Protein Lefty-2, Protein Lefty-A, TGFB4, TGF-beta 4, TGF-beta-4, Transforming Growth Factor Beta-4
Gene Symbols LEFTY2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Lefty-2 Phe78-Leu368 Accession # P57785
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320202
Test Specificity Detects mouse Lefty-2 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Lefty-A or recombinant mouse Lefty-1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.