Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Legumain/Asparaginyl Endopeptidase Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF2058SP
This item is not returnable.
View return policy
Description
Legumain/Asparaginyl Endopeptidase Polyclonal specifically detects Legumain/Asparaginyl Endopeptidase in Mouse samples. It is validated for Western Blot.Specifications
Legumain/Asparaginyl Endopeptidase | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
Sheep | |
Affinity Purified | |
RUO | |
5641 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL | |
O89017 | |
LGMN | |
Mouse myeloma cell line NS0-derived recombinant mouse Legumain Val18-Tyr435 Accession # O89017 | |
25 μg | |
Primary | |
Detects mouse Legumain in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction