Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Leptin/OB Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 13 publications
Supplier: R&D Systems AF498
This item is not returnable.
View return policy
Description
Leptin/OB Polyclonal specifically detects Leptin/OB in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
Leptin/OB | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
Goat | |
Affinity Purified | |
RUO | |
3952 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Western Blot, Immunohistochemistry, Neutralization, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.05-0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Q544U0 | |
LEP | |
E. coli-derived recombinant mouse Leptin/OB Val22-Cys167 Accession # Q544U0 | |
100 μg | |
Primary | |
Detects mouse Leptin/OB in ELISAs and Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction