Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Leptin R Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 18 publications
Supplier: R&D Systems AF497
This item is not returnable.
View return policy
Description
Leptin R Polyclonal specifically detects Leptin R in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
Leptin R | |
Polyclonal | |
Unconjugated | |
Q3US58 | |
LEPR | |
Mouse myeloma cell line NS0-derived recombinant mouse Leptin R Ala20-Gly839 Accession # Q3US58 | |
100 μg | |
Primary | |
Detects mouse Leptin R in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human Leptin R is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
Goat | |
Affinity Purified | |
RUO | |
3953 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction