Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Leptin R Biotinylated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. BAF497
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF497 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF497 Supplier R&D Systems Supplier No. BAF497

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 14 publications

Leptin R Polyclonal specifically detects Leptin R in Human∣Mouse∣Transgenic Mouse samples. It is validated for Bioassay∣Cell Selection∣ELISA Detection (Matched Antibody Pair)∣Flow Cytomery∣Flow Cytometry∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣In vivo Assay∣MACS∣Western Blot.

Specifications

Antigen Leptin R
Applications Bioassay, Cell Selection, ELISA Detection (Matched Antibody Pair), Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), In vivo Assay, Magnetic Cell Separation, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q3US58
Gene Alias B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295
Gene Symbols LEPR
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Leptin R Ala20-Gly839 Accession # Q3US58
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3953
Test Specificity Detects mouse Leptin R in ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Transgenic Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.