Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Lgr5/GPR49 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
$221.50 - $420.50
Specifications
Antigen | Lgr5/GPR49 |
---|---|
Clone | 803420 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.1 ug/10^6 cells |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB8240R025
![]() |
R&D Systems
FAB8240R025UG |
25 μg |
Each for $221.50
|
|
|||||
FAB8240R100
![]() |
R&D Systems
FAB8240R100UG |
100 μg |
Each for $420.50
|
|
|||||
Description
Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Mouse samples. It is validated for Flow Cytometry.Specifications
Lgr5/GPR49 | |
0.2 mg/mL | |
Flow Cytometry | |
Alexa Fluor 647 | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
LGR5 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse Lgr5/GPR49 in direct ELISAs. Stains mouse Lgr5 transfected cells but not irrelevant transfectants in flow cytometry. This clone, also known as “RD20”, has been found to detect an epitope in C-terminal LRR cap of LGR5 (Ref. 1). |
803420 | |
Flow Cytometry 0.1 ug/10^6 cells | |
Monoclonal | |
Rat | |
Mouse | |
Q9Z1P4 | |
8549 | |
E. coli-derived recombinant mouse Lgr5/GPR49 Lys375-Pro547 Accession # Q9Z1P4 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title