Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Lgr5/GPR49 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
$290.50 - $497.00
Specifications
Antigen | Lgr5/GPR49 |
---|---|
Clone | 889901 |
Dilution | Flow Cytometry 0.25-1 μg/106 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FB82401A025
![]() |
R&D Systems
FAB82401A025UG |
25 μg |
Each for $290.50
|
|
|||||
FB82401A100
![]() |
R&D Systems
FAB82401A100UG |
100 μg |
Each for $497.00
|
|
|||||
Description
Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Mouse samples. It is validated for Flow Cytometry.Specifications
Lgr5/GPR49 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Monoclonal | |
Supernatant | |
Mouse | |
Q9Z1P4 | |
8549.0 | |
Mouse Lgr5/GPR49 N-terminal peptide, Ala21-Cys34, Accession No. Q9Z1P4 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
889901 | |
Flow Cytometry | |
APC | |
Rat | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
LGR5 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse Lgr5/GPR49 in direct ELISAs. Stains mouse Lgr5/GPR49 transfected cells but not irrelevant transfectants in Flow Cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title