Learn More
Mouse Lgr5/GPR49 PE-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Lgr5/GPR49 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 803420 |
| Conjugate | R-PE |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | Q9Z1P4 |
| Gene Alias | FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 |
| Gene Symbols | LGR5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.